Learn More
Eppendorf™ Rotor for MiniSpin™ / MiniSpin™ Plus
Catalog No. 05414173 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
12 x 1.5/2.0 mL tubes
Catalog No. Capacity
05-414-173 12 x 1.5/2.0 mL tubes
1 options
This item is not returnable. View return policy
Catalog No. 05-414-173 Supplier Eppendorf™ Supplier No. 022668498
Add to Cart
Add to Cart
This item is not returnable. View return policy
Eppendorf Rotors for MiniSpin/MiniSpin plus, precision for mini centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile adaptors and buckets for efficient sample processing in a compact format.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
TRUSTED_SUSTAINABILITY
Specifications
Description F-45-12-11 - Includes rotor lid, rotor nut
Type Fixed Angle Rotor
Capacity 12 x 1.5/2.0 mL tubes
Max. RCF 14,100 x g
Max. Speed 14,500 rpm
Max. Tube Diameter 11 mm
For Use With (Equipment) MiniSpin™ / MiniSpin™ plus
Angle 45°
Includes Rotor Lid, rotor nut
No. of Places 12