Learn More
Eppendorf™ Rotor for Vacufuge Plus
Catalog No. 05414175 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
36 x 3.0/5.0 mL Flat-bottom Tubes
Catalog No. Capacity
05-414-175 36 x 3.0/5.0 mL Flat-bottom Tubes
1 options
This item is not returnable. View return policy
Catalog No. 05-414-175 Supplier Eppendorf™ Supplier No. 022822128
Add to Cart
Add to Cart
This item is not returnable. View return policy
Eppendorf Vacufuge Plus-efficiency meets precision. Tailor your concentration processes with a max speed of 1,700 rpm. Accommodate diverse samples with a 48 x 1.5/2.0 mL capacity. Customize your setup with various accessories for optimal sample concentration.
  • A wide range of fixed-angle rotors and the microplate rotor are available to accommodate a variety of tubes, plates, and applications.
  • Increase capacity with a spacer to stack some fixed-angle rotors.
  • Precision engineered for optimal sample concentration.
TRUSTED_SUSTAINABILITY
Specifications
Description F-45-36-15
Type Rotor
Capacity 36 x 3.0/5.0 mL Flat-bottom Tubes
Max. Tube Diameter 48 mm
For Use With (Equipment) Vacufuge Plus Vacuum Concentrators
No. of Places 36