Learn More
Eppendorf™ Rotor for Centrifuge 5420
Catalog No. 05413929 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
24 x 1.5/2.0 mL Tubes
Catalog No. Capacity
05-413-929 24 x 1.5/2.0 mL Tubes
1 options
Catalog No. 05-413-929 Supplier Eppendorf™ Supplier No. 5495400001
Add to Cart
Add to Cart
Eppendorf Rotors for Centrifuge 5420, precision for diverse applications. Choose from various capacities and speeds to tailor your centrifuge setup. Accessorize with adaptors and buckets, ensuring optimal sample processing in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
TRUSTED_SUSTAINABILITY
Specifications
Description FA-24x2, Aerosol-tight - Includes rotor lid
Material Anodized Aluminum
Type Rotor
Capacity 24 x 1.5/2.0 mL Tubes
Max. RCF 21,300 x g
Max. Speed 15,060 rpm
For Use With (Equipment) Centrifuge 5420
Includes Rotor Lid