Learn More
Sigma Aldrich Fine Chemicals Biosciences Rotor for EppendorfR 5804 EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP58207400001EA
Encompass
the S-4-104 rotor is not compatible with the 5804/5804R centrifuge. It can only be used with the 5810/5810R centrifuge
Catalog No. 50-290-8736
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.