Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Rotor Stand

Catalog No. 05414155 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Centrifuge 5804/5804 R/5810/5810 R
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-414-155 Centrifuge 5804/5804 R/5810/5810 R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-414-155 Supplier Eppendorf™ Supplier No. 022639021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rotor Stand for use with Centrifuge 5804/5804 R/5810/5810 R.

  • Centrifuge Stand
  • Fits all rotors rotors for Centrifuge 5804/5804 R/5810/5810 R
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Centrifuge 5804/5804 R/5810/5810 R
Disposable Reusable
Color Gray
Product Type Rotor Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.