Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian™ CP-14R, Chemical Resistant Rough Finish 14 mil Butyl Gloves
SDP

Catalog No. 19321125
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-321-125 Small
19-321-124 X-Small
19-321-126 Medium
19-321-127 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-321-125 Supplier Guardian™ Supplier No. 51602
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium weight butyl gloves with fitted hand styling and a rough finish texture offer protection against a wide range of hazardous chemicals.

  • Butyl material resists oxidation, ozone corrosion and abrasions
  • Reusable and easy to clean
  • Fitted hand style with rough finish texture
  • Protects against wide range of hazardous chemicals
  • 14 mil thickness, 14 in. length
  • Made in the USA

Specifications

Certifications/Compliance ASTM D412
Certifications/Compliance (Europe) CE
Color Black
Hand Specific
Latex Free Yes
Material Butyl
Powdered Powdered
Size Small
Thickness (English) 14 mil
Thickness (Metric) 0.35 mm
For Use With (Application) Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones
Glove Cuff Style Gauntlet/Rolled
Grip Design Rough
Lining Unlined
Manufacturing/QMS Audit Standards ISO 9001
Model 51602
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.