Learn More
IKA™ Flat Bottom Block
SDP
Catalog No. 01183261 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Catalog No. Capacity (Metric)
01-183-261 250 mL
1 options
This item is not returnable. View return policy
Catalog No. 01-183-261 Supplier IKA™ Supplier No. 25000633
Add to Cart
Add to Cart
This item is not returnable. View return policy
Compatible with several IKA stirrers.
  • Flat bottom wells
  • Volume Range: 8 to 250 mL
  • Number of Holes Available: 1, 4, 9 or 16
Specifications
Capacity (Metric) 250 mL
Includes Square carrier
Shape Round Bottom
Type Essay Block