Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Round Coverslips
SDPSCI_ED

Catalog No. S000847 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric):
12 mm
15 mm
18 mm
20 mm
25 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S000847 18 mm
S000845 12 mm
S000846 15 mm
S000848 20 mm
S000844 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Choking Hazard
Catalog No. S000847 Supplier United Scientific™ Supplier No. SQCVRSP18
Only null left
Add to Cart
Add to Cart
Choking Hazard

The Round Cover Slips are manufactured from soda lime glass.

  • Excellent resistance to chemicals
  • Ideal for neural tissue culture and hard-to-attach cells
  • Thickness ranges from 0.13 to 0.17 mm.
  • Avalible in multiple sizes

Specifications

Product Type Coverslip
Shape Circle
Material Glass
Length (Metric) 18 mm
Width (Metric) 18 mm
Thickness (Metric) 0.13 to 0.17 mm
Diameter (Metric) 18 mm
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.