Learn More
STOGGLES Round Lilac Dimmers
Supplier:  STOGGLES 154
Encompass_Preferred
Our Dimmers Lens are a UV-responsive lens that transition from light to dark based off of light exposure. Our light responsive lenses are anti-fog coated and shatter proof ensuring superior clarity and quality.
Catalog No. NC2388211
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.