Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ II DL247 Unisex Short Length Lab Jackets
SDP

Catalog No. 22412369
Change view
Click to view available options
Color:
Black
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-369 White Large
22-412-359 White X-Small
22-412-361 White Small
22-412-367 White Medium
22-412-372 White X-Large
22-412-377 White 3X-Large
22-412-363 Royal Blue X-Small
22-412-362 Royal Blue Small
22-412-364 Royal Blue Medium
22-412-370 Royal Blue Large
22-412-373 Royal Blue X-Large
22-412-375 Royal Blue 2X-Large
22-412-365 Royal Blue 3X-Large
22-412-360 Black X-Small
22-412-366 Black Small
22-412-368 Black Medium
22-412-371 Black Large
22-412-374 Black X-Large
22-412-376 Black 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Catalog No. 22-412-369 Supplier DenLine™ Uniforms Supplier No. DL247LWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lab coat provides AAMI PB70 Level 1 fluid resistance and offers protection from aerosols, blood, and all aqueous liquids.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Dental
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • AAMI PB70 Level 1 Protection
  • Provides wearers 5 ft. 2 in. and shorter frontal coverage to knees for OSHA 29 CFR 1910 compliance
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • Overall Air Permeability 45 CFM (ASTM D737)
  • Round neck, no knit ribbing to soil, fray or wear out prematurely
  • Stain resistant fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Lint free fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 Pockets, Stainless snap closures
  • Cuffs for overgloving
  • Bleach safe all colors, choice of 4 colors white, ceil blue, royal, and black
  • Autoclavable Fabric, 225 commercial wash tested

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polyester, Carbon
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Static, Stains
Style Short
No. of Pockets 3
Collar Type Round
Latex Free Yes
Size (English) Chest 42 to 44 in.
Size (English) Hip 40 to 42 in.
Size (Metric) Chest 107 to 112 cm
Size (Metric) Hip 102 to 107 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 1 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Length (English) 31.5 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 39.5 in.
Length (Metric) 80 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 45 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.