Learn More
DenLine™ Uniforms Protection Plus™ II DL247 Unisex Short Length Lab Jackets

Description
- Primary Applications:
- Anatomic Pathology
- Clinical Pathology
- Clinical Laboratory
- Phlebotomy
- Specimen Collection
- Blood Collection, Testing
- Manufacturing
- Dental
- Human and Animal Research
- Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
- AAMI PB70 Level 1 Protection
- Provides wearers 5 ft. 2 in. and shorter frontal coverage to knees for OSHA 29 CFR 1910 compliance
- Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
- Overall Air Permeability 45 CFM (ASTM D737)
- Round neck, no knit ribbing to soil, fray or wear out prematurely
- Stain resistant fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
- Anti-static fabric prevents false instrument readings, damage to computers
- Lint free fabric prevents sample contamination, ideal for molecular testing
- Durable construction, 3 Pockets, Stainless snap closures
- Cuffs for overgloving
- Bleach safe all colors, choice of 4 colors white, ceil blue, royal, and black
- Autoclavable Fabric, 225 commercial wash tested
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | Polyester, Carbon |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Fluids, Static, Stains |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.