Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Rous Sarcoma Virus (RAV-1) Avian Retrovirus, ATCC VR-334

Supplier:  ATCC VR334

Encompass

ATCC Rous sarcoma virus (VR-334) is an avian retrovirus preparation comprising a defective transforming virus (Bryan RSV) together with a replication-competent helper virus, RAV-1. It is antigenically identical to RAV-1, cross-reacts with antibodies to other avian leukosis viruses, and is supplied as a frozen preparation for research use. The material is propagated in chicken embryo cells and requires deep-freeze storage.

  • Strain: RAV-1 mixture including a defective transforming virus.
  • Host/propagation: chicken embryo cells.
  • Form: frozen viral preparation.
  • Storage: store at -70 °C or colder; stable at about -60 °C, not stable at -20 °C.
  • Intended use: research applications involving avian retroviruses and transformation studies.
  • Unit size: not specified by manufacturer.

Catalog No. NC3901771


May include imposed supplier surcharges.

Due to product restrictions, please to purchase or view availability for this product.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.