Learn More
Surgical Design Royaltek™ Stainless Steel Surgical Scalpel Blades
Catalog No. 22079684 Shop All Surgical Design Products
Change view
Click to view available options
Size:
10
11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-079-684 11
22-079-683 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-079-684 Supplier Surgical Design Supplier No. BLADE#11S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Surgical Design™ Royaltek™ Stainless Steel Surgical Scalpel Blades are made from sharp, long lasting surgical stainless steel. 10 series blades fit 3, 3L, 3 Graduated, 5B, 7, 9 B3, and B3L scalpel handles. 20 series blades fit 4, 4L, 4 Graduated and 6B.
  • Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade stainless-steel blades
  • Individually sealed
  • Peelable packing
  • Sterile
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
For Use With (Application) Chest Drain Incisions, opening Coronary Arteries and the Aorta and removing Calcifications in the Aortic or Mitral Valves
Size 11
For Use With (Equipment) Fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L.
Description Strong pointed Tip ideal for Stab Inscision
Sterility Sterile