Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rp-8-Br-cGMPS, cGMP Blocker, Purity >98%, 500 μg

Catalog No. 502705398
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5398 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5398 Supplier Abcam Supplier No. AB145204500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent cGMP blocker with >98% purity, designed for competitive inhibition studies.

  • cGMP blocker
  • Molecular weight: 462.15 Da
  • Purity: >98%
  • Competitively inhibits relaxation elicited by 8-Br-cGMP in isolated rabbit aorta contracted by phenylephrine at 30 μM

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule cGMP
Buffer 100% Rp-8-Br-cGMPS
Content And Storage Store at -20°C.
Shipping Condition Ambient
Formulation 100% Rp-8-Br-cGMPS
Quantity 500 μg
For Use With (Application) cGMP blocking
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.