Learn More
Tocris Bioscience™ Rp-8-pCPT-cGMPS sodium
Description
Specifications
Specifications
| Purity | >99% |
| Description | Rp-8-pCPT-cGMPS sodium |
| Molecular Weight (g/mol) | 525.86 |
| Product Type | Rp-8-pCPT-cGMPS sodium |
| Quantity | 1 mg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.