Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPA2 Monoclonal Antibody (9H8)
GREENER_CHOICE

Catalog No. MA126418 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA126418 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA126418 Supplier Invitrogen™ Supplier No. MA126418
  • $746.50
    $595.65 / Each of 1

    Save $150.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Recommended positive controls: Tonsil.

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9H8
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Rpa2
Gene Accession No. P15927
Gene Alias 30-kDa protein; AA409079; AI325195; AU020965; HSSB; hypothetical protein; I79_004200; p32; p32-subunit of replication protein A; p34; REPA2; Replication factor A protein 2; replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2; replication protein A2, 32kDa; replication protein A4; RFA; RF-A protein 2; Rf-A2; RP21C; RP4-547C9.3; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4; RpLP1; RpP2
Gene Symbols Rpa2
Host Species Mouse
Immunogen BALB/C mice were injected with purified human RPA protein.
Purification Method IgG fraction
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human, Monkey
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.