Learn More
Invitrogen™ RPAIN Monoclonal Antibody (OTI4F7)

Description
RPAIN Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RPAIN |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPAIN |
| Gene Accession No. | Q86UA6 |
| Gene Alias | 2400006N03Rik; hRIP; nuclear transporter; RAP interaction protein; RAP interaction protein alpha; RGD1308492; RIP; RPA interacting protein; RPA interacting protein delta 2; RPA interacting protein epsilon; RPAIN; RPA-interacting protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.