Learn More
Cayman Chemical RPalmItoyl 1methyl Ethno 5mg
SDP
Supplier:  Cayman Chemical 903535 MG
Encompass_Preferred
A PROTAC that drives Cdk9 degradation selectively induces degradation of Cdk9 over Cdk1 Cdk2 and Cdk7 in MOLT-4 cells at 250 nM inhibits HCMV replication in infected HFFs (EC50s 0.025 UM) inhibits viral replication in murine CMV-infected MEFs (EC50 0.22 UM)
Catalog No. 50-289-4255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.