Learn More
LABSYSTEMS DIAGNOSTICS OY Rapid detection of Helicobacter pylori antibodies
Supplier:  LABSYSTEMS DIAGNOSTICS OY 3018200
Encompass_Preferred
Biocard™ Helicobacter pylori IgG is a quick and sensitive home test to detect Helicobacter pylori antibodies in the blood. It is used as an aid in the diagnosis of a Helicobacter pylori infection, which may be connected with peptic ulcer disease, inflammation of gastric or duodenal mucosa and with increased risk of stomach cancer. The test requires only 1 drop of blood from the fingertip, and it can be performed and evaluated in 5-10 minutes
Catalog No. NC2865506