Learn More
Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Specifications
Specifications
| Antigen | RPIA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | RPIA |
| Gene Accession No. | P47968 |
| Gene Alias | Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD |
| Gene Symbols | RPIA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
