Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPIA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544350 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544350 Supplier Invitrogen™ Supplier No. PA544350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MQRPGPFSTL YGRVLAPLPG RAGGAASGGG GNSWDLPGSH VRLPGRAQSG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPIA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RPIA
Gene Accession No. P49247
Gene Alias Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD
Gene Symbols RPIA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RPIA (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22934
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.