Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL36A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567462 Supplier Invitrogen™ Supplier No. PA567462
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVNVPKTRRTF CKKCGKHQPH KVTQYKKGKD SLYAQG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

RPL36A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosol; endoplasmic reticulum; plasma membrane; ribosome; structural constituent of ribosome; translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL36A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPL36A
Gene Accession No. P83881
Gene Alias 60S ribosomal protein L36a; 60S ribosomal protein L36-A; 60S ribosomal protein L44; Cell growth-inhibiting gene 15 protein; cell migration-inducing gene 6 protein; dJ164F3.3 (ribosomal protein L44); GIG15; L36A; L39; L44L; L44-like ribosomal protein; Large ribosomal subunit protein eL36-A; Large ribosomal subunit protein eL42; MIG6; ribosomal 60S subunit protein L36A; ribosomal protein L36A; ribosomal protein L44; RPL36A; RPL39A; RPL39B; Rpl44; YL39; YM9646.06; YMR194W
Gene Symbols RPL36A
Host Species Rabbit
Immunogen Recombinant Human RPL36A. Recombinant protein control fragment (Product #RP-105671).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.