Learn More
Description
- Store at 10 to 25°C away from light
- Not classified as hazardous by current legislation, does not contain harmful substances in concentrations of ≥1%
- Research Use Only (RUO)
Specifications
Specifications
| Form | Agar |
| Format | 90 mm Plate |
| Manufacturing Origin | Italy |
| Product Type | Antifungal Medium |
| Sterility | Non-sterile |
| Target Organism Class | Fungi |
| Test Type | Fungal Testing |
| Quantity | 1 Ea. |
| Species | Yeast, Saccharomyces cerevisiae, Filamentous Fungi |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
