Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 2.0 g/L NaHCO3 (6 x 500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0416500X6
Encompass_Preferred
PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 2.0 g/L NaHCO3. QUANTITY: 6 x 500 ml. MANUFACTURED BY: Pan-Biotech (GERMANY). BICARBONATE: With 2.0 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with L-glutamine. HEPES: Without HEPES. LIQUID/POWDER: Liquid. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2°C - +8°C. SUMMARY: RPMI 1640, also known as RPMI Medium, was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells. We offer a variety of RPMI media to meet the needs of your specific applications.
Catalog No. 50-238-8624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.