Learn More
Invitrogen™ RPP40 Polyclonal Antibody

Description
Immunogen sequence: LSLNLDSKKY ERISWSFKEK KPLKFDFLLA WHKTGSEEST MMSYFSKYQI QEHQPKVALS TLRDLQCPVL QSSELEGTPE V.
Specifications
Specifications
| Antigen | RPP40 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.58 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RPP40 |
| Gene Accession No. | O75818, Q5BK64, Q8R1F9 |
| Gene Alias | bA428J1.3; D8Bwg1265e; ribonuclease P (40 kD); ribonuclease P 40 subunit; ribonuclease P 40kDa subunit; ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40 subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P/MRP subunit p40; ribonuclease P1; RNase P subunit 1; RNaseP protein p40; Rnasep1; RPP40 |
| Gene Symbols | RPP40 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.