Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS11 Antibody [Alexa Fluor™ Plus 680], Novus Biologicals™
SDP

Catalog No. RD10124686
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10124686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10124686 Supplier R&D Systems Supplier No. NBP222288AFP680

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

RPS11 Polyclonal antibody specifically detects RPS11 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)

Specifications

Antigen RPS11
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor Plus 680
Formulation 50mM Sodium Borate
Gene Alias ribosomal protein S11,40S ribosomal protein S11
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Ribosomal Protein S11 using the numbering given in entry NP_001006.1 (GeneID 6205).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6205
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.