Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116787 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116787 Supplier Invitrogen™ Supplier No. PA5116787
  • $587.00
    $489.65 / Each of 1

    Save $97.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: HepG2, Rat liver Immunogen sequence: MDTSRVQPIK LARVTKVLGR TGSQGQCTQV RVEFMDDTSR SIIRNVKGPV REGDVLTLLE SEREARRLR.

TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS28
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RPS28
Gene Accession No. P62857, P62858, P62859
Gene Alias 40S ribosomal protein S28; DBA15; fb07a10; ribosomal protein S28; rps28; S28; Small ribosomal subunit protein eS28; wu:fb07a10; wu:fb94f01; zgc:73367
Gene Symbols RPS28
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-69 of human RPS28 (NP_0010221).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54127, 6234, 691531
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.