Learn More
Invitrogen™ RPS4X Polyclonal Antibody

Description
RPS4X Polyclonal Antibody detects endogenous levels of RPS4X protein.
Specifications
Specifications
| Antigen | RPS4X |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPS4X |
| Gene Accession No. | P62701, P62702, P62703 |
| Gene Alias | 40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4 |
| Gene Symbols | RPS4X |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.