Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS6KL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555690 Supplier Invitrogen™ Supplier No. PA555690
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQAHVYLEQI RNRVALGVPD MTKRDYLVDA ATQIRLALER DVSEDYEAAF NHYQNGVDVL LRGIHVDPNK ERRE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.

RPS6KL1 (ribosomal protein S6 kinase-like 1) is a 549 amino acid protein that belongs to the Ser/Thr protein kinase family, S6 kinase subfamily and protein kinase superfamily. Existing as three alternatively spliced isoforms, RPS6KL1 contains one MIT domain and a protein kinase domain. The gene encoding RPS6KL1 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS6KL1
Gene Accession No. Q9Y6S9
Gene Alias A830084F09Rik; AW046440; I79_009419; ribosomal protein S6 kinase like 1; ribosomal protein S6 kinase-like 1; Rps6kl1; RSKL2
Gene Symbols RPS6KL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS6KL1. Recombinant protein control fragment (Product #RP-95246).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.