Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6KL1 Rabbit anti-Hamster, Human, Polyclonal, Proteintech
SDP

Catalog No. 501734524 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4524 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4524 Supplier Proteintech Group Inc Supplier No. 247571AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

RPS6KL1 (ribosomal protein S6 kinase-like 1) is a 549 amino acid protein that belongs to the Ser/Thr protein kinase family, S6 kinase subfamily and protein kinase superfamily. Existing as three alternatively spliced isoforms, RPS6KL1 contains one MIT domain and a protein kinase domain. The gene encoding RPS6KL1 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome.

Specifications

Antigen RPS6KL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RPS6KL1
Gene Accession No. Q9Y6S9
Gene Alias RPS6KL1
Gene Symbols RPS6KL1
Host Species Rabbit
Immunogen RPS6KL1 Fusion Protein Ag15099
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100758757, 83694
Target Species Hamster, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.