Learn More
Invitrogen™ RPUSD2 Polyclonal Antibody

Description
Peptide sequence: LKDNDFLRNT VHRHEPPVTA EPIRLLAENE DVVVVDKPSS IPVHPCGRFR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Specifications
Specifications
| Antigen | RPUSD2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RPUSD2 |
| Gene Accession No. | Q8IZ73 |
| Gene Alias | 4921503C21Rik; 9630001E10; BB231107; C15orf19; C18B11; C18B11 homolog (44.9kD); CG6187-like; FLJ31409; RGD1306147; RNA pseudouridine synthase domain containing 2; RNA pseudouridylate synthase domain containing 2; RNA pseudouridylate synthase domain-containing protein 2; Rpusd2 |
| Gene Symbols | RPUSD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.