Learn More
MilliporeSigma™ RQflex™ Reflectometer Accessories
Catalog No. M1169980001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Check set
Data transfer software package
Empty Cell
Recalibration set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1169980001 Data transfer software package
M1169570001 Check set
M1167270001 Empty Cell
M1169540001 Recalibration set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1169980001 Supplier MilliporeSigma™ Supplier No. 1.16998.0001
Accessories for RQflex™ and RQflex™ 10 plus reflectometers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Includes CD with software, 1 cable for RS232 interface and an adapter to connect the RS232 to USB interface of PC
Type Data transfer software package