Learn More
Enzo Life Sciences RR-src peptide (1mg). CAS: 81156-93-6
SDP
Supplier:  Enzo Life Sciences BMLP3080001
Encompass_Preferred
This tyrosine kinase substrate sequence, RRLIEDAEYAARG, was derived from the v-Src autophosphorylation site (original src sequence is IEDNEYTARQG). Km=5 mM in LSTRA cell extracts. Alternative name: v-src autophosphorylation site. Purity: ≥97%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1339
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.