Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGA/RRAGB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101819 Supplier Invitrogen™ Supplier No. PA5101819
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RRAGA/B.

Ras-homologous GTPases constitute a large family of signal transducers that alternate between an activated, GTP-binding state and an inactivated, GDP-binding state. These proteins represent cellular switches that are operated by GTP-exchange factors and factors that stimulate their intrinsic GTPase activity. All GTPases of the Ras superfamily have in common the presence of six conserved motifs involved in GTP/GDP binding, three of which are phosphate-/magnesium-binding sites (PM1-PM3) and three of which are guanine nucleotide-binding sites (G1-G3). Transcript variants encoding distinct isoforms have been identified. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGA/RRAGB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RRAGB
Gene Accession No. Q5VZM2, Q63486, Q63487, Q6NTA4, Q7L523, Q80X95
Gene Alias 1300010C19Rik; adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; bA465E19.1; FIP1; FIP-1; GTP-binding protein ragB; rag A; Rag B; Raga; Ragb; Ras related GTP binding A; Ras related GTP binding B; Ras-related GTP binding A; Ras-related GTP binding B; ras-related GTP-binding protein A; Ras-related GTP-binding protein B; Ras-related GTP-binding protein ragA; RGD:619804}; RGD:619805}; RRAGA; rraga {ECO:0000312; RragB; rragB {ECO:0000312
Gene Symbols RRAGA, RRAGB
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRAGA(Accession Q7L523), corresponding to amino acid residues N269-R313.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10325, 10670, 117043, 117044, 245670, 68441
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.