Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531659 Supplier Invitrogen™ Supplier No. PA531659
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (91%), Chicken (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RRP12 (Ribosomal RNA Processing 12 Homolog) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includebinding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RRP12
Gene Accession No. Q5JTH9
Gene Alias AA408556; AA536972; Kiaa0690; mKIAA0690; ribosomal RNA processing 12 homolog; ribosomal RNA processing 12 homolog (S. cerevisiae); Rrp12; RRP12-like protein
Gene Symbols RRP12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human RRP12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.