Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565456 Supplier Invitrogen™ Supplier No. PA565456
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYDQKIAEEEA KAKEEEGVPD EEGWVKVTRR GRRPVLPRTE AASLRVLERE RRKRS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

RRP7A (Ribosomal RNA Processing 7 Homolog A) is a Protein Coding gene. Diseases associated with RRP7A include Autosomal Dominant Non-Syndromic Intellectual Disability 1 and Autosomal Dominant Non-Syndromic Intellectual Disability. Among its related pathways are rRNA processing in the nucleus and cytosol and Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleotide binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP7A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RRP7A
Gene Accession No. Q9Y3A4
Gene Alias 1110014J01Rik; AA408146; BK126B4.3; CGI-96; CTA-126B4.5; gastric cancer antigen Zg14; Gastric cancer antigen Zg14 homolog; hypothetical protein LOC550241; Kheg1; kidney highly expressed protein 1; RGD1311547; ribosomal RNA processing 7 homolog; ribosomal RNA processing 7 homolog A; ribosomal RNA processing 7 homolog A (S. cerevisiae); ribosomal RNA-processing protein 7 homolog A; RRP7; RRP7A; zgc:110084
Gene Symbols RRP7A
Host Species Rabbit
Immunogen Recombinant Human RRP7A. Recombinant protein control fragment (Product #RP-106134).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27341
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.