Learn More
Cayman Chemical RS-1, 312756-74-4, 10 mg
SDP
Supplier:  Cayman Chemical 2103710
Encompass_Preferred
An activator of RAD51 (Kd = 48-107 for human RAD51); a potent enhancer of CRISPR-mediated genome editing
Catalog No. NC1429966
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.