Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RS 1

Catalog No. 581050
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
581050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 581050 Supplier Tocris Bioscience™ Supplier No. 5810/50
This item is not returnable. View return policy

High affinity RAD51 activator; also CRISPR enhancer

High affinity RAD51 activator (Kd range = 48-107 nM, value depends on cofactors present). Enhances homologous recombination activity of hRAD51. Enhances homology-directed repair ∽2 to 5-fold and increases CRISPR-mediated knock-in efficiencies in vitro and in vivo.

Specifications

Chemical Name or Material 4-Bromo-N-(4-bromophenyl)-3-[[(phenylmethyl)amino]sulfonyl]benzamide
CAS 312756-74-4
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Inhibitors
Molecular Formula C20H16Br2N2O3S
Formula Weight 524.23
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.