Learn More
Medchemexpress LLC 6H-isoquino[2,1-g][1,6]naphthyridine, 12-(ethylsulfonyl)-3-methoxy hydrochloride hemihydrate | 186002-54-0 | 99.5% | 410.06 g/mol | C19H28N2O3S·HCl·1/2H2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101338A10MG
Encompass_Preferred
RS-79948-197 hemihydrate is a research-grade α2-adrenoceptor antagonist supplied as the hydrochloride hemihydrate salt for in vitro pharmacology and receptor-binding studies. It is provided as a white to off-white solid with high purity and recommended sealed, refrigerated storage to preserve stability.
  • High purity (99.5%) suitable for research assays.
  • Hydrochloride hemihydrate form for improved stability.
  • White to off-white solid, easy to handle and weigh.
  • Stable when stored sealed at 4°C, with defined solvent storage conditions.
  • Established pharmacology as an α2-adrenoceptor antagonist for receptor-binding studies.
Catalog No. 50-002-11989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.