Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119394 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119394 200 μL
PIPA5121633 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119394 Supplier Invitrogen™ Supplier No. PA5119394
  • $536.00
    $457.65 / Each of 1

    Save $78.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RS1 Polyclonal Antibody for Western Blot

May be active in cell adhesion processes during retinal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene RS1
Gene Accession No. O15537
Gene Alias retinoschisin; retinoschisin 1; retinoschisis (X-linked, juvenile) 1 (human); retinoschisis 1 homolog; RS; RS1; Rs1h; tmgc1; X-linked juvenile retinoschisis protein; X-linked juvenile retinoschisis protein homolog; XLRS1
Gene Symbols RS1
Host Species Rabbit
Immunogen Recombinant protein Retinoschisin. The antigen corresponds to amino acid range 24-224 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106506914, 6247
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.