Learn More
RSAD2 Mouse anti-Human, Clone: OTI4D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505799

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Viperin is a 43kDa protein which is one of several hundred interferon inducible viral proteins. Elevated expression of viperin is induced by LPS and poly I:C through TLR dependent signaling pathways. Sendai virus mediated viperin expression is induced independently of TLR signaling pathways through RLRs - RIG-I and MAVs (Severa, 2006). Viperin has been shown to play an important role in interferon mediated anti-viral activity in a Hepatitis C virus model system (Helbig, 2011). Viperin is induced in lymphoid cells and dendritic cells (DCs) and highly induced in neutrophils and macrophages during acute LCMV infection in mice (Hinson, ). Viperin functions as a mediator of viral resistance and as a marker for interferon responsiveness making its detection and mechanism of action through specific antibody probes valuable for further studies.Specifications
RSAD2 | |
Monoclonal | |
Unconjugated | |
RSAD2 | |
2510004L01Rik, cig33, cig5, vig1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
91543 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WXG1 | |
RSAD2 | |
Full length human recombit protein of human RSAD2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.