Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSBN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544728 Supplier Invitrogen™ Supplier No. PA544728
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GGAVGPFKCV FVGEMAAQVG AVRVVRAVAA QEEPDKEGKE KPHAGVSPRG Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Histone demethylase that specifically demethylates dimethylated 'Lys-20' of histone H4 (H4K20me2), thereby modulating chromosome architecture. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSBN1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RSBN1
Gene Accession No. Q5VWQ0
Gene Alias C230004D03Rik; E330004N01; KDM9; Kiaa3002; Lysine-specific demethylase 9; mKIAA3002; RGD1307231; Rosbin; rosbin, round spermatid basic protein 1; round spermatid basic protein 1; Rsbn; Rsbn1; Rsbp
Gene Symbols RSBN1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RSBN1 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.