Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSL1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116393
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116393 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116393 Supplier Invitrogen™ Supplier No. PA5116393
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSL1D1 Polyclonal Antibody for Western Blot

Regulates cellular senescence through inhibition of PTEN translation. Acts as a pro-apoptotic regulator in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSL1D1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RSL1D1
Gene Accession No. O76021
Gene Alias 2410005K20Rik; Ac1158; C76439; CATX11; CATX-11; cellular senescence inhibited; cellular senescence inhibited gene protein; cellular senescence-inhibited gene protein; CSIG; DKFZP564M182; L12; MGC138433; MGC142259; pBK1; Protein PBK1; ribosomal L1 domain containing 1; ribosomal L1 domain-containing protein 1; RSL1D1; UTP30
Gene Symbols RSL1D1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 455-485 amino acids from human RSL1D1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.