Learn More
TARGETMOL CHEMICALS INC RSL3 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3646
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RSL3 (RSL3 1S) is a VDAC-independent ferroptosis activator with selectivity for tumor cells bearing oncogenic RAS. RSL3 is the inhibitor of the glutathione peroxidase 4 which can inhibit the cysteine/glutamate amino acid transporter system xc- that blocks GSH synthesis (IC50 100 nM). purity: 99%
Catalog No. 50-257-6897
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.