Learn More
ACROBiosystems Recombinant Protein;Human R-Spondin 1 / RSPO1 (21-146) Protein;HEK293;1MG;RS6,RSPO1,RSPO-1,CRISTIN3,CRISTIN-3,FLJ40906,RP11-566C13.1,RSPO,R-spondin-1,Rspondin-1,Rspondin1,Rspondin
Supplier:  ACROBiosystems RS6H42201MG
Encompass_Preferred
Recombinant Protein;Human R-Spondin 1 / RSPO1 (21-146) Protein;1MG;Human R-Spondin 1, His Tag (RS6-H4220) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession # AAI14967).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7721
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.