Learn More
Pyramex™ RSSH32 Series Winter Wear Pullover Sweatshirt
Catalog No. 19555347 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-347 5X-Large
19-555-343 Medium
19-555-342 Large
19-555-348 X-Large
19-555-344 2X-Large
19-555-345 3X-Large
19-555-346 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-555-347 Supplier Pyramex™ Supplier No. RSSH3210X5
Add to Cart
Add to Cart
This item is not returnable. View return policy
RSSH32 Series Winter Wear Pullover Sweatshirt is part of the Pyramex™ Safety Hi -Vis Winter Wear line.
  • 100% polyester 280 gsm fleece
  • Elastic knit at wrists and waist
  • 2 in. silver reflective material
  • Pull over type sweat shirt with hood
  • Mesh lined hood with adjustable pull cord
  • 1 large front pocket
Specifications
Size 5X-Large
Gender Unisex
Material Polyester Fleece
Color Hi-vis Lime
Neck Style Hooded
Seam Type Sewn
Sleeve Style Long Sleeve
Cuff Style Elastic Knit
Disposable Reusable
No. of Pockets 1
Certifications/Compliance ANSI/ISEA 107-2015; Type R - Class 3
Tape Type China Brand Flat Silver
Thickness 280 gsm