Learn More
Enzo Life Sciences RSSR (50mg). CAS: 123557-49-3
SDP
Supplier:  Enzo Life Sciences ALX430102M050
Encompass_Preferred
Cell permeable supersensitive spin label (detection limit varies from 10nM to 100nM) for quick detection of reduced thiols using ESR. Allows following the reactions of sulfhydryl groups with RSSR to form thiol spin label adducts, for the monitoring of intracellular redox states of glutathione and other thiols. Alternative name: bis-(2,2,5,5-Tetramethyl-3-imidazoline-1-oxyl-4-yl)disulfide, biradical. Purity: ≥97% (HPLC, GLC). Appearance: Off-white to yellow crystalline powder. Solubility: Soluble in DMSO, water, 100% ethanol or methanol; DMSO is recommended for stock solution. Long Term Storage: -20°C.
Catalog No. 50-200-9205
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.