Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543158 Supplier Invitrogen™ Supplier No. PA543158
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PIADQFQLGV SHVFEYIRSE TYKYLYGRHM QANPEPPKKN NDKSKKISRK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Sheep: 93%; Zebrafish: 93%.

RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation. This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSU1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RSU1
Gene Accession No. Q15404
Gene Alias cubilin; ras suppressor protein 1; etID12695; FLJ31034; LOC553276 protein; ras suppressor protein 1; ras suppressor protein 1 isoform 1; ras suppressor protein 1 variant 1; ras suppressor protein 1 variant 2; ras suppressor protein 1 variant 3; ras suppressor protein 1 variant 5; Ras suppressor protein 1-like protein; RSP1; RSP-1; rsu1; Rsu-1; RsuI; si:dz63m2.1; Unknown (protein for MGC:126965)
Gene Symbols RSU1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RSU1 (aa 221-270).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6251
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.