Learn More
Description
Specifications
Specifications
| Assay Range | 125 pg/mL |
| Assay Sensitivity | 125-8000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Line | Novus |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human respiratory syncytial virus (RSV) Fusion Protein. |
| Target Species | Virus |
| For Use With (Application) | Sandwich ELISA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
