Learn More
Sino Biological RSV RSV-F MAb 20UG 11049-R338-H-20
Supplier:  Sino Biological 11049R338H20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human respiratory syncytial virus (RSV) Fusion glycoprotein / RSV-F (Catalog#11049-V08B; P03420.1; Met1-Thr529) and was produced using recombinant antibody technology and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-9640
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.